CD34 Monoclonal Antibody (RAM34), eFluor™ 450, eBioscience™ - FAQs
CD34 Monoclonal Antibody (RAM34), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD34 Monoclonal Antibody (RAM34), eFluor™ 450, eBioscience™ - FAQs (48-0341-82, 48-0341-80)