CD34 Monoclonal Antibody (4H11), eFluor™ 450, eBioscience™ - FAQs
CD34 Monoclonal Antibody (4H11), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD34 Monoclonal Antibody (4H11), eFluor™ 450, eBioscience™ - FAQs (48-0349-41, 48-0349-42)