CD38 Monoclonal Antibody (90), eFluor™ 450, eBioscience™ - FAQs
CD38 Monoclonal Antibody (90), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD38 Monoclonal Antibody (90), eFluor™ 450, eBioscience™ - FAQs (48-0381-82, 48-0381-80)