CD38 Monoclonal Antibody (HIT2), eFluor™ 450, eBioscience™ - FAQs
CD38 Monoclonal Antibody (HIT2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD38 Monoclonal Antibody (HIT2), eFluor™ 450, eBioscience™ - FAQs (48-0389-42, 48-0389-41)