CD40 Monoclonal Antibody (HM40-3), eFluor™ 450, eBioscience™ - FAQs
CD40 Monoclonal Antibody (HM40-3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD40 Monoclonal Antibody (HM40-3), eFluor™ 450, eBioscience™ - FAQs (48-0402-82, 48-0402-80)