CD40 Monoclonal Antibody (5C3), eFluor™ 450, eBioscience™ - FAQs
CD40 Monoclonal Antibody (5C3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD40 Monoclonal Antibody (5C3), eFluor™ 450, eBioscience™ - FAQs (48-0409-41, 48-0409-42)