CD41a Monoclonal Antibody (HIP8), eFluor™ 450, eBioscience™ - FAQs
CD41a Monoclonal Antibody (HIP8), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD41a Monoclonal Antibody (HIP8), eFluor™ 450, eBioscience™ - FAQs (48-0419-41, 48-0419-42)