CD42a Monoclonal Antibody (GR-P), eFluor™ 450, eBioscience™ - FAQs
CD42a Monoclonal Antibody (GR-P), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD42a Monoclonal Antibody (GR-P), eFluor™ 450, eBioscience™ - FAQs (48-0428-42, 48-0428-41)