CD44 Monoclonal Antibody (IM7), eFluor™ 450, eBioscience™ - FAQs
CD44 Monoclonal Antibody (IM7), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD44 Monoclonal Antibody (IM7), eFluor™ 450, eBioscience™ - FAQs (48-0441-80, 48-0441-82)