CD45 Monoclonal Antibody (30-F11), eFluor™ 450, eBioscience™ - FAQs
CD45 Monoclonal Antibody (30-F11), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (30-F11), eFluor™ 450, eBioscience™ - FAQs (48-0451-80, 48-0451-82)