CD45.1 Monoclonal Antibody (A20), eFluor™ 450, eBioscience™ - FAQs
CD45.1 Monoclonal Antibody (A20), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45.1 Monoclonal Antibody (A20), eFluor™ 450, eBioscience™ - FAQs (48-0453-80, 48-0453-82)