CD45RO Monoclonal Antibody (UCHL1), eFluor™ 450, eBioscience™ - FAQs
CD45RO Monoclonal Antibody (UCHL1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45RO Monoclonal Antibody (UCHL1), eFluor™ 450, eBioscience™ - FAQs (48-0457-41, 48-0457-42)