CD45RA Monoclonal Antibody (HI100), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45RA Monoclonal Antibody (HI100), eFluor™ 450, eBioscience™ - FAQs (48-0458-41, 48-0458-42)