CD45 Monoclonal Antibody (HI30), eFluor™ 450, eBioscience™ - FAQs
CD45 Monoclonal Antibody (HI30), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (HI30), eFluor™ 450, eBioscience™ - FAQs (48-0459-41, 48-0459-42)