CD45 Monoclonal Antibody (OX1), eFluor™ 450, eBioscience™ - FAQs
CD45 Monoclonal Antibody (OX1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (OX1), eFluor™ 450, eBioscience™ - FAQs (48-0461-82, 48-0461-80)