CD48 Monoclonal Antibody (HM48-1), eFluor™ 450, eBioscience™ - FAQs
CD48 Monoclonal Antibody (HM48-1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD48 Monoclonal Antibody (HM48-1), eFluor™ 450, eBioscience™ - FAQs (48-0481-80, 48-0481-82)