CD56 (NCAM) Monoclonal Antibody (TULY56), eFluor™ 450, eBioscience™ - FAQs
CD56 (NCAM) Monoclonal Antibody (TULY56), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD56 (NCAM) Monoclonal Antibody (TULY56), eFluor™ 450, eBioscience™ - FAQs (48-0566-41, 48-0566-42)