CD57 Monoclonal Antibody (TB01 (TBO1)), eFluor™ 450, eBioscience™ - FAQs
CD57 Monoclonal Antibody (TB01 (TBO1)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD57 Monoclonal Antibody (TB01 (TBO1)), eFluor™ 450, eBioscience™ - FAQs (48-0577-41, 48-0577-42)