CD69 Monoclonal Antibody (H1.2F3), eFluor™ 450, eBioscience™ - FAQs
CD69 Monoclonal Antibody (H1.2F3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD69 Monoclonal Antibody (H1.2F3), eFluor™ 450, eBioscience™ - FAQs (48-0691-82, 48-0691-80)