CD73 Monoclonal Antibody (eBioTY/11.8 (TY/11.8)), eFluor™ 450, eBioscience™ - FAQs
CD73 Monoclonal Antibody (eBioTY/11.8 (TY/11.8)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD73 Monoclonal Antibody (eBioTY/11.8 (TY/11.8)), eFluor™ 450, eBioscience™ - FAQs (48-0731-80, 48-0731-82)