CD80 (B7-1) Monoclonal Antibody (16-10A1), eFluor™ 450, eBioscience™ - FAQs
CD80 (B7-1) Monoclonal Antibody (16-10A1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD80 (B7-1) Monoclonal Antibody (16-10A1), eFluor™ 450, eBioscience™ - FAQs (48-0801-82, 48-0801-80)