CD86 (B7-2) Monoclonal Antibody (GL1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD86 (B7-2) Monoclonal Antibody (GL1), eFluor™ 450, eBioscience™ - FAQs (48-0862-82, 48-0862-80)