CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), eFluor™ 450, eBioscience™ - FAQs
CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), eFluor™ 450, eBioscience™ - FAQs (48-0900-80, 48-0900-82)