CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), eFluor™ 450, eBioscience™ - FAQs
CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), eFluor™ 450, eBioscience™ - FAQs (48-0902-82, 48-0902-80)