CD94 Monoclonal Antibody (18d3), eFluor™ 450, eBioscience™ - FAQs
CD94 Monoclonal Antibody (18d3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD94 Monoclonal Antibody (18d3), eFluor™ 450, eBioscience™ - FAQs (48-0941-82)