CD95 (APO-1/Fas) Monoclonal Antibody (DX2), eFluor™ 450, eBioscience™ - FAQs
CD95 (APO-1/Fas) Monoclonal Antibody (DX2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD95 (APO-1/Fas) Monoclonal Antibody (DX2), eFluor™ 450, eBioscience™ - FAQs (48-0959-41, 48-0959-42)