CD103 (Integrin alpha E) Monoclonal Antibody (2E7), eFluor™ 450, eBioscience™ - FAQs
CD103 (Integrin alpha E) Monoclonal Antibody (2E7), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD103 (Integrin alpha E) Monoclonal Antibody (2E7), eFluor™ 450, eBioscience™ - FAQs (48-1031-82, 48-1031-80)