CD105 (Endoglin) Monoclonal Antibody (MJ7/18), eFluor™ 450, eBioscience™ - FAQs
CD105 (Endoglin) Monoclonal Antibody (MJ7/18), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD105 (Endoglin) Monoclonal Antibody (MJ7/18), eFluor™ 450, eBioscience™ - FAQs (48-1051-82, 48-1051-80)