CD105 (Endoglin) Monoclonal Antibody (SN6), eFluor™ 450, eBioscience™ - FAQs
CD105 (Endoglin) Monoclonal Antibody (SN6), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD105 (Endoglin) Monoclonal Antibody (SN6), eFluor™ 450, eBioscience™ - FAQs (48-1057-41, 48-1057-42)