CD106 (VCAM-1) Monoclonal Antibody (429), eFluor™ 450, eBioscience™ - FAQs
CD106 (VCAM-1) Monoclonal Antibody (429), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD106 (VCAM-1) Monoclonal Antibody (429), eFluor™ 450, eBioscience™ - FAQs (48-1061-80, 48-1061-82)