CD117 (c-Kit) Monoclonal Antibody (2B8), eFluor™ 450, eBioscience™ - FAQs
CD117 (c-Kit) Monoclonal Antibody (2B8), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD117 (c-Kit) Monoclonal Antibody (2B8), eFluor™ 450, eBioscience™ - FAQs (48-1171-82, 48-1171-80)