CD117 (c-Kit) Monoclonal Antibody (104D2), eFluor™ 450, eBioscience™ - FAQs
CD117 (c-Kit) Monoclonal Antibody (104D2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD117 (c-Kit) Monoclonal Antibody (104D2), eFluor™ 450, eBioscience™ - FAQs (48-1178-41, 48-1178-42)