CD123 Monoclonal Antibody (7G3), eFluor™ 450, eBioscience™ - FAQs
CD123 Monoclonal Antibody (7G3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD123 Monoclonal Antibody (7G3), eFluor™ 450, eBioscience™ - FAQs (48-1238-42)