CD123 Monoclonal Antibody (6H6), eFluor™ 450, eBioscience™ - FAQs
CD123 Monoclonal Antibody (6H6), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD123 Monoclonal Antibody (6H6), eFluor™ 450, eBioscience™ - FAQs (48-1239-42, 48-1239-41)