CD127 Monoclonal Antibody (A7R34), eFluor™ 450, eBioscience™ - FAQs
CD127 Monoclonal Antibody (A7R34), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD127 Monoclonal Antibody (A7R34), eFluor™ 450, eBioscience™ - FAQs (48-1271-80, 48-1271-82)