CD127 Monoclonal Antibody (eBioRDR5), eFluor™ 450, eBioscience™ - FAQs
CD127 Monoclonal Antibody (eBioRDR5), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD127 Monoclonal Antibody (eBioRDR5), eFluor™ 450, eBioscience™ - FAQs (48-1278-41, 48-1278-42)