CD146 Monoclonal Antibody (P1H12), eFluor™ 450, eBioscience™ - FAQs
CD146 Monoclonal Antibody (P1H12), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD146 Monoclonal Antibody (P1H12), eFluor™ 450, eBioscience™ - FAQs (48-1469-41, 48-1469-42)