CD150 Monoclonal Antibody (mShad150), eFluor™ 450, eBioscience™ - FAQs
CD150 Monoclonal Antibody (mShad150), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD150 Monoclonal Antibody (mShad150), eFluor™ 450, eBioscience™ - FAQs (48-1502-80, 48-1502-82)