CD154 (CD40 Ligand) Monoclonal Antibody (MR1), eFluor™ 450, eBioscience™ - FAQs
CD154 (CD40 Ligand) Monoclonal Antibody (MR1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD154 (CD40 Ligand) Monoclonal Antibody (MR1), eFluor™ 450, eBioscience™ - FAQs (48-1541-82, 48-1541-80)