CD154 (CD40 Ligand) Monoclonal Antibody (24-31), eFluor™ 450, eBioscience™ - FAQs
CD154 (CD40 Ligand) Monoclonal Antibody (24-31), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD154 (CD40 Ligand) Monoclonal Antibody (24-31), eFluor™ 450, eBioscience™ - FAQs (48-1548-41, 48-1548-42)