CD158a/h/g Monoclonal Antibody (HP-MA4), eFluor™ 450, eBioscience™ - FAQs
CD158a/h/g Monoclonal Antibody (HP-MA4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD158a/h/g Monoclonal Antibody (HP-MA4), eFluor™ 450, eBioscience™ - FAQs (48-1589-41)