CD161 Monoclonal Antibody (HP-3G10), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD161 Monoclonal Antibody (HP-3G10), eFluor™ 450, eBioscience™ - FAQs (48-1619-41, 48-1619-42)