CD197 (CCR7) Monoclonal Antibody (4B12), eFluor™ 450, eBioscience™ - FAQs
CD197 (CCR7) Monoclonal Antibody (4B12), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD197 (CCR7) Monoclonal Antibody (4B12), eFluor™ 450, eBioscience™ - FAQs (48-1971-80, 48-1971-82)