CD223 (LAG-3) Monoclonal Antibody (3DS223H), eFluor™ 450, eBioscience™ - FAQs
CD223 (LAG-3) Monoclonal Antibody (3DS223H), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD223 (LAG-3) Monoclonal Antibody (3DS223H), eFluor™ 450, eBioscience™ - FAQs (48-2239-41, 48-2239-42)