CD304 (Neuropilin-1) Monoclonal Antibody (3DS304M), eFluor™ 450, eBioscience™ - FAQs
CD304 (Neuropilin-1) Monoclonal Antibody (3DS304M), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD304 (Neuropilin-1) Monoclonal Antibody (3DS304M), eFluor™ 450, eBioscience™ - FAQs (48-3041-80, 48-3041-82)