CD335 (NKp46) Monoclonal Antibody (29A1.4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD335 (NKp46) Monoclonal Antibody (29A1.4), eFluor™ 450, eBioscience™ - FAQs (48-3351-82, 48-3351-80)