CD335 (NKp46) Monoclonal Antibody (9E2), eFluor™ 450, eBioscience™ - FAQs
CD335 (NKp46) Monoclonal Antibody (9E2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD335 (NKp46) Monoclonal Antibody (9E2), eFluor™ 450, eBioscience™ - FAQs (48-3359-42, 48-3359-41)