CD336 (NKp44) Monoclonal Antibody (44.189), eFluor™ 450, eBioscience™ - FAQs
CD336 (NKp44) Monoclonal Antibody (44.189), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD336 (NKp44) Monoclonal Antibody (44.189), eFluor™ 450, eBioscience™ - FAQs (48-3369-41, 48-3369-42)