CD337 (NKp30) Monoclonal Antibody (AF29-4D12), eFluor™ 450, eBioscience™ - FAQs
CD337 (NKp30) Monoclonal Antibody (AF29-4D12), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD337 (NKp30) Monoclonal Antibody (AF29-4D12), eFluor™ 450, eBioscience™ - FAQs (48-3379-42)