Arginase 1 Monoclonal Antibody (A1exF5), eFluor™ 450, eBioscience™ - FAQs
Arginase 1 Monoclonal Antibody (A1exF5), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Arginase 1 Monoclonal Antibody (A1exF5), eFluor™ 450, eBioscience™ - FAQs (48-3697-80, 48-3697-82)